Assay Details
Target Gene Details
Entrez Gene ID: | 10161 |
Gene Name: | lysophosphatidic acid receptor 6 |
Gene Aliases: |
ARWH1, HYPT8, LAH3, P2RY5, P2Y5 |
Location: |
Chr.13:48411045-48444704 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LPAR6 | NM_001162497.1 | 5 | 584 | NP_001155969.1 |
| NM_001162498.1 | 1 | 535 | NP_001155970.1 | |
| NM_005767.5 | 7 | 1066 | NP_005758.2 | |
| AK122856.1 | 5 | 584 | ||
| AK123059.1 | 1 | 131 | ||
| BC040850.1 | 1 | 118 | AAH40850.1 | |
| BC045651.1 | 7 | 1067 | AAH45651.1 | |
| BC070295.1 | ||||
| BF950635.1 | ||||
| BF951153.1 | ||||
| BX537392.1 | 1 | 131 | CAD97634.1 | |
| BX537438.1 | 1 | 131 | CAD97680.1 | |
| BX537445.1 | 1 | 131 | CAD97687.1 | |
| BX647109.1 | 1 | 131 |
Target Gene Details
Entrez Gene ID: | 5925 |
Gene Name: | RB transcriptional corepressor 1 |
Gene Aliases: |
OSRC, PPP1R130, RB, p105-Rb, pRb, pp110 |
Location: |
Chr.13:48303747-48481890 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RB1 | NM_000321.2 | NP_000312.2 | ||
| XM_011535171.2 | XP_011533473.1 | |||
| AB208788.1 | BAD92025.1 | |||
| AK291258.1 | ||||
| AK299179.1 | ||||
| AK300284.1 | ||||
| AK307730.1 | ||||
| BC039060.1 | AAH39060.1 | |||
| BC040540.1 | AAH40540.1 | |||
| L41870.1 | AAB59465.1 | |||
| M15400.1 | AAA69807.1 | |||
| M28419.1 | AAA69808.1 | |||
| M33647.1 | AAA69806.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1649n100 | Chr.13:48397374 - 48422859 on Build GRCh38 | Gain |
|
| nsv1122816 | Chr.13:48395084 - 48417263 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map