Assay Details
Target Gene Details
Entrez Gene ID: | 54680 |
Gene Name: | zinc finger HIT-type containing 6 |
Gene Aliases: |
BCD1, C1orf181, NY-BR-75 |
Location: |
Chr.1:85649423-85708447 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNHIT6 | NM_001170670.1 | 11 | 2188 | NP_001164141.1 |
| NM_017953.3 | 10 | 2305 | NP_060423.3 | |
| AF308296.1 | 10 | 1951 | AAG48263.1 | |
| AK000736.1 | 10 | 1677 | BAA91349.1 | |
| AK000767.1 | 10 | 2214 | BAA91371.1 | |
| AL442074.1 | 10 | 2290 | CAC09440.1 | |
| BC026236.1 | 10 | 2292 | AAH26236.1 | |
| BC110898.1 | 10 | 2223 | AAI10899.1 |
More Information
Set Membership: |
|

Genomic Map