Assay Details
Target Gene Details
Entrez Gene ID: | 10389 |
Gene Name: | sex comb on midleg-like 2 (Drosophila) |
Gene Aliases: |
- |
Location: |
Chr.X:18239313-18355089 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCML2 | NM_006089.2 | 15 | 3008 | NP_006080.1 |
| NR_033717.1 | ||||
| XM_006724458.1 | 14 | 2820 | XP_006724521.1 | |
| XM_017029221.1 | 14 | 2890 | XP_016884710.1 | |
| AK022824.1 | 3 | 1256 | ||
| AL833937.1 | CAD38792.1 | |||
| BC051913.1 | AAH51913.1 | |||
| BC064617.1 | 15 | 2965 | AAH64617.1 | |
| Y18004.1 | 15 | 2942 | CAB38943.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2231e212 | Chr.X:18217755 - 18242206 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map