Assay Details
Target Gene Details
Entrez Gene ID: | 337973 |
Gene Name: | keratin associated protein 19-6 |
Gene Aliases: |
KAP19.6 |
Location: |
Chr.21:30541535-30541864 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRTAP19-6 | NM_001303120.1 | NP_001290049.1 | ||
| NM_181612.3 | NP_853643.1 | |||
| AB096947.1 | BAE46362.1 | |||
| AB096948.1 | BAE46363.1 | |||
| BC101813.1 | ||||
| BC101815.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057467 | Chr.21:30318003 - 30993921 on Build GRCh38 | Gain |
|
| nsv587354 | Chr.21:29978351 - 30620545 on Build GRCh38 | Gain |
|

Genomic Map