Assay Details
Target Gene Details
Entrez Gene ID: | 55731 |
Gene Name: | family with sequence similarity 222 member B |
Gene Aliases: |
C17orf63 |
Location: |
Chr.17:28755978-28855036 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM222B | NM_001077498.2 | 3 | 3287 | NP_001070966.1 |
| NM_001288631.1 | 4 | 3458 | NP_001275560.1 | |
| NM_001288632.1 | 5 | 3369 | NP_001275561.1 | |
| NM_001288633.1 | 4 | 3363 | NP_001275562.1 | |
| NM_001288634.1 | 3 | 3356 | NP_001275563.1 | |
| NM_001288635.1 | 4 | 3335 | NP_001275564.1 | |
| NM_001288636.1 | 6 | 3359 | NP_001275565.1 | |
| NM_001288637.1 | 5 | 3325 | NP_001275566.1 | |
| NM_001288638.1 | 5 | 3257 | NP_001275567.1 | |
| NM_001288639.1 | 4 | 3209 | NP_001275568.1 | |
| NM_001288640.1 | 3 | 3175 | NP_001275569.1 | |
| NM_018182.3 | 4 | 3321 | NP_060652.2 | |
| XM_011524986.2 | 3 | 3252 | XP_011523288.1 | |
| XM_011524987.1 | 4 | 3087 | XP_011523289.1 | |
| XM_011524988.2 | 3 | 3303 | XP_011523290.1 | |
| XM_017024831.1 | 4 | 3411 | XP_016880320.1 | |
| XM_017024832.1 | 3 | 3676 | XP_016880321.1 | |
| XM_017024833.1 | 3 | 3216 | XP_016880322.1 | |
| XM_017024834.1 | 3 | 3414 | XP_016880323.1 | |
| XM_017024835.1 | 4 | 3613 | XP_016880324.1 | |
| AK001562.1 | 4 | 3272 | BAA91757.1 | |
| CB156081.1 | 1 | 548 |

Genomic Map