Assay Details
Target Gene Details
Entrez Gene ID: | 79576 |
Gene Name: | NFKB activating protein |
Gene Aliases: |
- |
Location: |
Chr.X:119925050-119943772 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKAP | NM_024528.3 | 1 | 456 | NP_078804.2 |
| AK026279.1 | 1 | 456 | BAB15428.1 | |
| AL832118.1 | 3 | 397 | ||
| AY388958.1 | 1 | 456 | AAQ90402.1 | |
| BC015354.1 | 1 | 320 | AAH15354.1 | |
| BC016783.1 | 1 | 324 | ||
| BC018968.1 | 1 | 427 | ||
| BC032442.1 | 1 | 314 | AAH32442.1 | |
| BC071686.1 | 1 | 435 | AAH71686.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758888 | Chr.X:119823476 - 120195131 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map