Assay Details
Target Gene Details
Entrez Gene ID: | 6746 |
Gene Name: | signal sequence receptor subunit 2 |
Gene Aliases: |
HSD25, TLAP, TRAP-BETA, TRAPB |
Location: |
Chr.1:156009048-156020992 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSR2 | NM_003145.3 | NP_003136.1 | ||
| XM_011509923.2 | XP_011508225.1 | |||
| AK092473.1 | ||||
| AK222600.1 | BAD96320.1 | |||
| AK300678.1 | ||||
| AK313821.1 | ||||
| AY251536.1 | AAP20059.1 | |||
| BC000341.2 | ||||
| BP196192.1 | ||||
| CR456973.1 | CAG33254.1 | |||
| D37991.1 | BAA07206.1 | |||
| X74104.1 | CAA52207.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509513 | Chr.1:155932322 - 156178790 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map