Assay Details
Target Gene Details
Entrez Gene ID: | 6400 |
Gene Name: | SEL1L ERAD E3 ligase adaptor subunit |
Gene Aliases: |
PRO1063, SEL1-LIKE, SEL1L1 |
Location: |
Chr.14:81471547-81533864 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEL1L | NM_005065.5 | 21 | 7354 | NP_005056.3 |
| AB020335.1 | 21 | 7282 | BAA87904.1 | |
| AK075511.1 | ||||
| CA447094.1 | 1 | 589 | ||
| CB986742.1 | 1 | 585 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3635076 | Chr.14:80715257 - 81672373 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map