Assay Details
Target Gene Details
Entrez Gene ID: | 10713 |
Gene Name: | ubiquitin specific peptidase 39 |
Gene Aliases: |
65K, CGI-21, HSPC332, SAD1, SNRNP65 |
Location: |
Chr.2:85602842-85649284 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP39 | NM_001256725.1 | NP_001243654.1 | ||
| NM_001256726.1 | NP_001243655.1 | |||
| NM_001256727.1 | NP_001243656.1 | |||
| NM_001256728.1 | NP_001243657.1 | |||
| NM_006590.3 | NP_006581.2 | |||
| NR_046347.1 | ||||
| XM_006711922.1 | XP_006711985.1 | |||
| XM_006711923.2 | XP_006711986.1 | |||
| XM_011532487.1 | XP_011530789.1 | |||
| XM_011532488.1 | XP_011530790.1 | |||
| XM_017003182.1 | XP_016858671.1 | |||
| AF132955.1 | AAD27730.1 | |||
| AF353989.1 | AAK49524.1 | |||
| AK001047.1 | ||||
| AK056334.1 | ||||
| AK129872.1 | ||||
| AK222852.1 | BAD96572.1 | |||
| AK289451.1 | ||||
| AK295257.1 | ||||
| AK302236.1 | ||||
| AK309485.1 | ||||
| AL519827.3 | ||||
| BC001384.2 | AAH01384.2 | |||
| BC067273.1 | AAH67273.1 | |||
| BX462623.2 | ||||
| DA548921.1 | ||||
| DB124294.1 | ||||
| DB444135.1 |
More Information
Set Membership: |
|

Genomic Map