Assay Details
Target Gene Details
Entrez Gene ID: | 1983 |
Gene Name: | eukaryotic translation initiation factor 5 |
Gene Aliases: |
EIF-5, EIF-5A |
Location: |
Chr.14:103334002-103345025 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF5 | NM_001969.4 | NP_001960.2 | ||
| NM_183004.4 | NP_892116.2 | |||
| AK026933.1 | BAB15593.1 | |||
| AK293712.1 | ||||
| AL080102.1 | CAB45711.1 | |||
| BC007728.2 | AAH07728.1 | |||
| BC032866.2 | AAH32866.1 | |||
| BX429104.2 | ||||
| BX537367.1 | CAD97610.1 | |||
| CR456712.1 | CAG32993.1 | |||
| CR627002.1 | 1 | 271 | ||
| U49436.1 | AAC50572.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9168 | Chr.14:103065392 - 103446969 on Build GRCh38 | Loss |
|
| nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map