Assay Details
Target Gene Details
Entrez Gene ID: | 149420 |
Gene Name: | PDLIM1 interacting kinase 1 like |
Gene Aliases: |
CLIK1L, STK35L2 |
Location: |
Chr.1:26109406-26125548 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDIK1L | NM_001243532.1 | 3 | 2542 | NP_001230461.1 |
| NM_001243533.1 | 4 | 2616 | NP_001230462.1 | |
| NM_152835.4 | 3 | 2425 | NP_690048.1 | |
| XM_005245743.4 | 4 | 2685 | XP_005245800.1 | |
| XM_006710381.3 | 2 | 3999 | XP_006710444.1 | |
| XM_011540792.2 | 2 | 2096 | XP_011539094.1 | |
| AF411102.1 | 4 | 2556 | AAN03661.1 | |
| AL137407.1 | 1 | 733 | ||
| AL834290.1 | 1 | 1871 | CAD38964.1 | |
| BC028713.1 | 4 | 2558 | AAH28713.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv511129 | Chr.1:26123610 - 26141503 on Build GRCh38 | Loss |
|
| nsv950339 | Chr.1:26121910 - 26127709 on Build GRCh38 | Duplication |
|
| nsv819393 | Chr.1:26123804 - 26124506 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map