Assay Details
Target Gene Details
Entrez Gene ID: | 3888 |
Gene Name: | keratin 82 |
Gene Aliases: |
HB2, Hb-2, KRTHB2 |
Location: |
Chr.12:52393951-52406392 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT82 | NM_033033.3 | NP_149022.3 | ||
| AJ628419.1 | CAF31523.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832410 | Chr.12:52321596 - 52506980 on Build GRCh38 | Gain |
|
| nsv558928 | Chr.12:52401803 - 52437285 on Build GRCh38 | Loss |
|
| nsv832411 | Chr.12:52347803 - 52494080 on Build GRCh38 | Loss |
|
| nsv832412 | Chr.12:52390206 - 52545452 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map