Assay Details
Target Gene Details
Entrez Gene ID: | 59349 |
Gene Name: | kelch like family member 12 |
Gene Aliases: |
C3IP1, DKIR |
Location: |
Chr.1:202891096-202928644 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHL12 | NM_001303051.1 | NP_001289980.1 | ||
| NM_001303109.1 | NP_001290038.1 | |||
| NM_021633.3 | NP_067646.1 | |||
| XM_011509835.1 | XP_011508137.1 | |||
| XM_011509836.1 | XP_011508138.1 | |||
| XM_011509837.1 | XP_011508139.1 | |||
| XM_017001995.1 | XP_016857484.1 | |||
| AF190900.1 | AAG17175.1 | |||
| AF306687.1 | ||||
| AF306689.1 | ||||
| AK024412.1 | BAB14914.1 | |||
| AK027656.1 | BAB55271.1 | |||
| AK223072.1 | BAD96792.1 | |||
| AK301791.1 | ||||
| BC003183.1 | AAH03183.1 | |||
| BC004175.2 | AAH04175.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832359 | Chr.1:202739288 - 202918866 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map