Assay Details
Target Gene Details
Entrez Gene ID: | 2237 |
Gene Name: | flap structure-specific endonuclease 1 |
Gene Aliases: |
FEN-1, MF1, RAD2 |
Location: |
Chr.11:61792637-61797244 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FEN1 | NM_004111.5 | 1 | 57 | NP_004102.1 |
| BP244711.1 | 1 | 37 | ||
| CN290841.1 | 1 | 57 | ||
| X76771.1 | CAA54166.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528697 | Chr.11:61690385 - 61919561 on Build GRCh38 | Loss |
|
| nsv343 | Chr.11:61770698 - 61802486 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs143036029,rs148183125] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map