Assay Details
Target Gene Details
Entrez Gene ID: | 127254 |
Gene Name: | glutamate rich 3 |
Gene Aliases: |
C1orf173 |
Location: |
Chr.1:74568111-74674343 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERICH3 | NM_001002912.4 | 15 | 5693 | NP_001002912.4 |
| XM_017000274.1 | 14 | 6352 | XP_016855763.1 | |
| XM_017000276.1 | 4 | 4121 | XP_016855765.1 | |
| XM_017000277.1 | 3 | 4079 | XP_016855766.1 | |
| XM_017000278.1 | 4 | 4094 | XP_016855767.1 | |
| XM_017000279.1 | 4 | 4548 | XP_016855768.1 | |
| XM_017000280.1 | 3 | 4489 | XP_016855769.1 | |
| XM_017000281.1 | 2 | 4845 | XP_016855770.1 | |
| XM_017000282.1 | 3 | 4052 | XP_016855771.1 | |
| XM_017000283.1 | 3 | 4506 | XP_016855772.1 | |
| XM_017000284.1 | 2 | 4300 | XP_016855773.1 | |
| XM_017000285.1 | 2 | 4447 | XP_016855774.1 | |
| AL833250.1 | 2 | 1001 | ||
| AL834414.1 | 2 | 2760 | CAD39076.1 | |
| CR749349.1 | 3 | 3616 | CAH18202.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830248 | Chr.1:74496707 - 74678774 on Build GRCh38 | Loss |
|
| nsv1002115 | Chr.1:72379872 - 74933684 on Build GRCh38 | Gain |
|

Genomic Map