Assay Details
Target Gene Details
Entrez Gene ID: | 3073 |
Gene Name: | hexosaminidase subunit alpha |
Gene Aliases: |
TSD |
Location: |
Chr.15:72343435-72376473 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HEXA | NM_000520.5 | 1 | 97 | NP_000511.2 |
| NM_001318825.1 | 1 | 97 | NP_001305754.1 | |
| NR_134869.1 | 1 | 97 | ||
| CR627386.1 | 1 | 97 | CAH10482.1 |
Target Gene Details
Entrez Gene ID: | 80072 |
Gene Name: | HEXA antisense RNA 1 |
Gene Aliases: |
C15orf34 |
Location: |
Chr.15:72376113-72378788 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HEXA-AS1 | NR_027262.1 | 1 | 265 | |
| AK023377.1 | BAB14550.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521971 | Chr.15:72374083 - 72386397 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map