Assay Details
Target Gene Details
Entrez Gene ID: | 7105 |
Gene Name: | tetraspanin 6 |
Gene Aliases: |
T245, TM4SF6, TSPAN-6 |
Location: |
Chr.X:100627108-100637104 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPAN6 | NM_001278740.1 | 8 | 2255 | NP_001265669.1 |
| NM_001278741.1 | 8 | 2025 | NP_001265670.1 | |
| NM_001278742.1 | 7 | 1917 | NP_001265671.1 | |
| NM_001278743.1 | 7 | 1941 | NP_001265672.1 | |
| NM_003270.3 | 8 | 1963 | NP_003261.1 | |
| AB209876.1 | 8 | 1922 | BAD93113.1 | |
| AF043906.1 | 8 | 1867 | AAC64257.1 |
More Information
Set Membership: |
|

Genomic Map