Assay Details
Target Gene Details
Entrez Gene ID: | 4990 |
Gene Name: | SIX homeobox 6 |
Gene Aliases: |
MCOPCT2, ODRMD, OPTX2, Six9 |
Location: |
Chr.14:60509220-60511807 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIX6 | NM_007374.2 | 1 | 608 | NP_031400.2 |
| AJ011785.1 | 1 | 503 | CAA09773.1 | |
| BC065831.1 | 1 | 608 | AAH65831.1 | |
| BC069413.1 | 1 | 512 | AAH69413.2 | |
| BC087852.1 | 1 | 586 | AAH87852.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564880 | Chr.14:60397865 - 60916029 on Build GRCh38 | Gain |
|
| nsv564881 | Chr.14:60457291 - 61377535 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map