Assay Details
Target Gene Details
Entrez Gene ID: | 170850 |
Gene Name: | potassium voltage-gated channel modifier subfamily G member 3 |
Gene Aliases: |
KV10.1, KV6.3 |
Location: |
Chr.2:42442017-42494097 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNG3 | NM_133329.5 | 2 | 3561 | NP_579875.1 |
| NM_172344.2 | 2 | 3528 | NP_758847.1 | |
| AF454547.1 | 2 | 3409 | AAM93548.1 | |
| AF454548.1 | 2 | 3442 | AAM93549.1 | |
| BC071558.1 | 2 | 3441 | AAH71558.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1118e212 | Chr.2:42436818 - 42448184 on Build GRCh38 | Loss |
|
| nsv524679 | Chr.2:42350030 - 42576225 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map