Assay Details
Target Gene Details
Entrez Gene ID: | 65267 |
Gene Name: | WNK lysine deficient protein kinase 3 |
Gene Aliases: |
PRKWNK3 |
Location: |
Chr.X:54192823-54358695 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WNK3 | NM_001002838.3 | 23 | 5722 | NP_001002838.1 |
| NM_020922.4 | 24 | 5893 | NP_065973.2 | |
| XM_006724593.3 | 23 | 5660 | XP_006724656.1 | |
| XM_011530802.2 | 24 | 5690 | XP_011529104.1 | |
| XM_017029741.1 | 24 | 5832 | XP_016885230.1 | |
| XM_017029742.1 | 24 | 5909 | XP_016885231.1 | |
| XM_017029743.1 | 24 | 5824 | XP_016885232.1 | |
| XM_017029744.1 | 24 | 5726 | XP_016885233.1 | |
| XM_017029745.1 | 23 | 5801 | XP_016885234.1 | |
| AB046786.1 | 17 | 3994 | BAB13392.1 | |
| AJ409088.2 | CAC32455.2 | |||
| AY082340.1 | AAL99253.1 | |||
| AY352048.1 | 24 | 5893 | AAR89465.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion |
|
| nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map