Assay Details
Target Gene Details
Entrez Gene ID: | 55578 |
Gene Name: | SPT20 homolog, SAGA complex component |
Gene Aliases: |
C13, C13orf19, FAM48A, FP757, P38IP, SPT20 |
Location: |
Chr.13:37009312-37059714 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892330 | Chr.13:36864962 - 37227904 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map