Assay Details
Target Gene Details
Entrez Gene ID: | 84992 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class Y |
Gene Aliases: |
HPMRS6, PIG-Y |
Location: |
Chr.4:88520978-88523801 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGY | NM_001042616.2 | 2 | 430 | NP_001036081.1 |
| AB206972.1 | 2 | 402 | BAE44507.1 | |
| BC007876.2 | 2 | 402 | ||
| BI548853.1 | 2 | 435 |
Target Gene Details
Entrez Gene ID: | 100996939 |
Gene Name: | PIGY upstream reading frame |
Gene Aliases: |
PREY |
Location: |
Chr.4:88520978-88523801 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYURF | NM_032906.4 | 2 | 430 | NP_116295.1 |
| AK312082.1 | ||||
| BG706896.1 | 2 | 435 | ||
| CV812938.1 | 2 | 366 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv980229 | Chr.4:88521147 - 88521989 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map