Assay Details
Target Gene Details
Entrez Gene ID: | 54443 |
Gene Name: | anillin actin binding protein |
Gene Aliases: |
FSGS8, Scraps, scra |
Location: |
Chr.7:36389803-36453791 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANLN | NM_001284301.2 | 9 | 1826 | NP_001271230.1 |
| NM_001284302.2 | 9 | 1823 | NP_001271231.1 | |
| NM_018685.4 | 10 | 1937 | NP_061155.2 | |
| XM_006715746.1 | 10 | 1934 | XP_006715809.1 | |
| XM_006715747.3 | 9 | 1823 | XP_006715810.1 | |
| XM_017012354.1 | 10 | 1934 | XP_016867843.1 | |
| XM_017012355.1 | 10 | 1934 | XP_016867844.1 | |
| XM_017012356.1 | 9 | 1823 | XP_016867845.1 | |
| AF273437.1 | 10 | 1872 | AAF75796.1 | |
| AK001472.1 | 5 | 604 | BAA91711.1 | |
| AK023208.1 | BAB14463.1 | |||
| AK291254.1 | 9 | 1755 | ||
| AK299803.1 | 10 | 1639 | ||
| BC070066.1 | 9 | 1808 | AAH70066.1 | |
| CR936650.1 | 7 | 1027 | CAI56788.1 |
More Information
Set Membership: |
|

Genomic Map