Assay Details
Target Gene Details
Entrez Gene ID: | 401466 |
Gene Name: | chromosome 8 open reading frame 59 |
Gene Aliases: |
- |
Location: |
Chr.8:85214053-85220422 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C8orf59 | NM_001099670.2 | 5 | 868 | NP_001093140.1 |
| NM_001099671.2 | 5 | 865 | NP_001093141.1 | |
| NM_001099672.2 | 5 | 781 | NP_001093142.1 | |
| NM_001099673.2 | 4 | 740 | NP_001093143.1 | |
| NM_001293320.1 | 3 | 623 | NP_001280249.1 | |
| BC146983.1 | 4 | 685 | ||
| BC146988.1 | 4 | 685 | ||
| BC157825.1 | 4 | 685 |
Target Gene Details
Entrez Gene ID: | 1875 |
Gene Name: | E2F transcription factor 5 |
Gene Aliases: |
E2F-5 |
Location: |
Chr.8:85177384-85214518 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E2F5 | NM_001083588.1 | 8 | 1456 | NP_001077057.1 |
| NM_001083589.1 | 8 | 1310 | NP_001077058.1 | |
| NM_001951.3 | 8 | 1459 | NP_001942.2 | |
| AB209185.1 | 8 | 2427 | BAD92422.1 | |
| DB534924.2 | 1 | 439 | ||
| U15642.1 | AAC50120.1 | |||
| U31556.1 | 8 | 1457 | AAB00179.1 | |
| X86097.1 | 8 | 1452 | CAA60051.1 | |
| Z78409.1 | 8 | 1456 | CAB01634.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483126 | Chr.8:83687766 - 85887771 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map