Assay Details
Target Gene Details
Entrez Gene ID: | 7782 |
Gene Name: | solute carrier family 30 member 4 |
Gene Aliases: |
ZNT4, znT-4 |
Location: |
Chr.15:45479613-45523685 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC30A4 | NM_013309.5 | 8 | 1692 | NP_037441.2 |
| XM_011521997.2 | 8 | 2196 | XP_011520299.1 | |
| XM_017022560.1 | 7 | 1498 | XP_016878049.1 | |
| AF025409.1 | 7 | 1380 | AAB82561.1 | |
| AK290874.1 | ||||
| BC026089.2 | AAH26089.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469721 | Chr.15:45316683 - 45495804 on Build GRCh38 | Gain |
|
| esv3892684 | Chr.15:45307172 - 46327637 on Build GRCh38 | Gain |
|
| esv3569553 | Chr.15:45366990 - 46080626 on Build GRCh38 | Gain |
|
| nsv1160292 | Chr.15:45351161 - 46175480 on Build GRCh38 | Duplication |
|
| nsv1523 | Chr.15:45457702 - 45491627 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map