Assay Details
Target Gene Details
Entrez Gene ID: | 84684 |
Gene Name: | INSM transcriptional repressor 2 |
Gene Aliases: |
IA-6, IA6, mlt1 |
Location: |
Chr.14:35534042-35537054 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INSM2 | NM_032594.3 | 1 | 2061 | NP_115983.3 |
| AB037912.1 | 1 | 1936 | BAB69042.1 | |
| AF260323.1 | 1 | 2061 | AAK49393.1 | |
| AK096639.1 | 1 | 1936 | BAC04830.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039975 | Chr.14:35432355 - 35536457 on Build GRCh38 | Gain |
|
| nsv564340 | Chr.14:35534869 - 35536453 on Build GRCh38 | Loss |
|
| nsv564333 | Chr.14:35534103 - 35536453 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map