Assay Details
Target Gene Details
Entrez Gene ID: | 89849 |
Gene Name: | autophagy related 16 like 2 |
Gene Aliases: |
ATG16B, WDR80 |
Location: |
Chr.11:72814406-72843740 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATG16L2 | XM_005274376.4 | XP_005274433.1 | ||
| BQ929430.1 |
Target Gene Details
Entrez Gene ID: | 9873 |
Gene Name: | FCH and double SH3 domains 2 |
Gene Aliases: |
NWK, SH3MD3 |
Location: |
Chr.11:72836743-73142682 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCHSD2 | NM_014824.2 | 20 | 3659 | NP_055639.2 |
| XM_011545409.1 | 20 | 3541 | XP_011543711.1 | |
| XM_011545410.2 | 20 | 3474 | XP_011543712.1 | |
| XM_017018632.1 | 20 | 3718 | XP_016874121.1 | |
| AB018312.1 | 19 | 3509 | BAA34489.2 | |
| AK128381.1 | 1 | 2932 | ||
| AL080236.1 | 1 | 559 | ||
| AL133567.1 | 15 | 3027 | CAB63720.1 | |
| BC017751.1 | 8 | 2209 | AAH17751.1 |
More Information
Set Membership: |
|

Genomic Map