Assay Details
Target Gene Details
Entrez Gene ID: | 81494 |
Gene Name: | complement factor H related 5 |
Gene Aliases: |
CFHL5, CFHR5D, FHR-5, FHR5 |
Location: |
Chr.1:196975022-197009725 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFHR5 | NM_030787.3 | NP_110414.1 | ||
| XM_011510020.2 | XP_011508322.1 | |||
| AF295327.1 | AAK15619.1 | |||
| AK314026.1 | ||||
| BC026282.1 | ||||
| BC111773.1 | AAI11774.1 | |||
| DA638244.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv12e55 | Chr.1:196762018 - 197042447 on Build GRCh38 | Gain |
|
| nsv469599 | Chr.1:196827346 - 196986100 on Build GRCh38 | Loss |
|
| nsv428290 | Chr.1:196621707 - 196992672 on Build GRCh38 | Gain+Loss |
|
| nsv471614 | Chr.1:196700381 - 196986100 on Build GRCh38 | Gain+Loss |
|
| nsv528460 | Chr.1:196573437 - 197477572 on Build GRCh38 | Gain |
|
| nsv8702 | Chr.1:196974722 - 196985445 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map