Assay Details
Target Gene Details
Entrez Gene ID: | 57407 |
Gene Name: | NmrA-like family domain containing 1 |
Gene Aliases: |
HSCARG, SDR48A1 |
Location: |
Chr.16:4461677-4476320 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NMRAL1 | NM_001305141.1 | NP_001292070.1 | ||
| NM_001305142.1 | NP_001292071.1 | |||
| NM_020677.4 | NP_065728.1 | |||
| XM_006720905.3 | XP_006720968.1 | |||
| XM_006720906.2 | XP_006720969.1 | |||
| XM_011522567.2 | XP_011520869.1 | |||
| XM_017023485.1 | XP_016878974.1 | |||
| XM_017023486.1 | XP_016878975.1 | |||
| XM_017023487.1 | XP_016878976.1 | |||
| XM_017023488.1 | XP_016878977.1 | |||
| XM_017023489.1 | XP_016878978.1 | |||
| XM_017023490.1 | XP_016878979.1 | |||
| XM_017023491.1 | XP_016878980.1 | |||
| XM_017023492.1 | XP_016878981.1 | |||
| AF225419.1 | AAG09721.1 | |||
| AK098784.1 | ||||
| AW246763.1 | ||||
| BC002927.2 | AAH02927.1 | |||
| BC007364.2 | AAH07364.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160332 | Chr.16:4367643 - 4486554 on Build GRCh38 | Duplication |
|
| nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
|
| esv3584924 | Chr.16:4330628 - 4469258 on Build GRCh38 | Gain |
|
| esv23732 | Chr.16:4340813 - 4476244 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map