Assay Details
Target Gene Details
Entrez Gene ID: | 55466 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member A4 |
Gene Aliases: |
MST104, MSTP104, PRO1472 |
Location: |
Chr.15:78264145-78282196 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJA4 | NM_001130182.1 | 1 | 166 | NP_001123654.1 |
| NM_018602.3 | 2 | 305 | NP_061072.3 | |
| XM_005254535.3 | 2 | 78 | XP_005254592.1 | |
| XM_006720601.1 | 2 | 80 | XP_006720664.1 | |
| AK096616.1 | 1 | 168 | BAC04828.1 | |
| AK098079.1 | 2 | 296 | BAC05229.1 | |
| BC021720.2 | 2 | 305 | AAH21720.1 | |
| BX648710.1 | 2 | 187 | CAH10558.1 | |
| DB047940.1 | 2 | 305 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv570200 | Chr.15:78241768 - 78273499 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map