Assay Details
Target Gene Details
Entrez Gene ID: | 54936 |
Gene Name: | ADP-ribosylhydrolase like 2 |
Gene Aliases: |
ARH3 |
Location: |
Chr.1:36088852-36093932 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADPRHL2 | NM_017825.2 | NP_060295.1 | ||
| XM_011541636.2 | XP_011539938.1 | |||
| AF212236.1 | AAK14922.1 | |||
| AJ427295.1 | CAD20316.1 | |||
| AK000453.1 | BAA91174.1 | |||
| AK223037.1 | BAD96757.1 | |||
| AK295283.1 | ||||
| AK316349.1 | ||||
| BC014169.2 | AAH14169.1 | |||
| CR457237.1 | CAG33518.1 | |||
| DC365461.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834691 | Chr.1:36059137 - 36238540 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map