Assay Details
Target Gene Details
Entrez Gene ID: | 51763 |
Gene Name: | inositol polyphosphate-5-phosphatase K |
Gene Aliases: |
PPS, SKIP |
Location: |
Chr.17:1494575-1516888 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INPP5K | NM_001135642.1 | 14 | 3100 | NP_001129114.1 |
| NM_016532.3 | 12 | 2710 | NP_057616.2 | |
| NM_130766.2 | 13 | 2941 | NP_570122.1 | |
| XM_005256683.2 | 12 | 2550 | XP_005256740.1 | |
| XM_005256685.1 | 11 | 2331 | XP_005256742.1 | |
| XM_005256686.1 | 12 | 2679 | XP_005256743.1 | |
| XM_011523934.1 | 14 | 2723 | XP_011522236.1 | |
| XM_011523936.2 | 12 | 2560 | XP_011522238.1 | |
| XM_017024755.1 | 13 | 2781 | XP_016880244.1 | |
| XM_017024756.1 | 14 | 2858 | XP_016880245.1 | |
| XM_017024757.1 | 13 | 2756 | XP_016880246.1 | |
| XM_017024758.1 | 12 | 2490 | XP_016880247.1 | |
| XM_017024759.1 | 11 | 2341 | XP_016880248.1 | |
| AB036829.1 | 12 | 2495 | BAA92340.1 | |
| AB036830.1 | 13 | 2724 | BAA92341.1 | |
| BM510617.1 | 1 | 287 | ||
| U45973.1 | 9 | 1964 | AAB03214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833334 | Chr.17:1411327 - 1582381 on Build GRCh38 | Loss |
|
| esv3582474 | Chr.17:1494346 - 1505407 on Build GRCh38 | Loss |
|
| nsv952098 | Chr.17:1464507 - 1515106 on Build GRCh38 | Deletion |
|
| nsv1057782 | Chr.17:1351585 - 1512616 on Build GRCh38 | Gain |
|
| dgv877e212 | Chr.17:1494279 - 1500654 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map