Assay Details
Target Gene Details
Entrez Gene ID: | 785 |
Gene Name: | calcium voltage-gated channel auxiliary subunit beta 4 |
Gene Aliases: |
CAB4, CACNLB4, EA5, EIG9, EJM, EJM4, EJM6 |
Location: |
Chr.2:151832771-152099988 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNB4 | NM_000726.3 | 14 | 3040 | NP_000717.2 |
| NM_001005746.2 | 14 | 2990 | NP_001005746.1 | |
| NM_001005747.2 | 13 | 3128 | NP_001005747.1 | |
| NM_001145798.1 | 13 | 2854 | NP_001139270.1 | |
| NM_001320722.1 | 14 | 3288 | NP_001307651.1 | |
| XM_011511795.2 | 14 | 4757 | XP_011510097.1 | |
| XM_011511796.2 | 13 | 3711 | XP_011510098.1 | |
| XM_011511797.2 | 11 | 3760 | XP_011510099.1 | |
| XM_017004880.1 | 11 | 2852 | XP_016860369.1 | |
| XM_017004881.1 | 10 | 2920 | XP_016860370.1 | |
| XM_017004882.1 | 11 | 4245 | XP_016860371.1 | |
| XM_017004883.1 | 15 | 4579 | XP_016860372.1 | |
| XM_017004884.1 | 13 | 4413 | XP_016860373.1 | |
| XM_017004885.1 | 12 | 4253 | XP_016860374.1 | |
| XM_017004886.1 | 5 | 2333 | XP_016860375.1 | |
| BX484858.1 | 1 | 498 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34659 | Chr.2:150858366 - 152860232 on Build GRCh38 | Loss |
|
| esv3592922 | Chr.2:151571476 - 153829714 on Build GRCh38 | Loss |
|
| esv1010582 | Chr.2:151835724 - 151845268 on Build GRCh38 | Deletion |
|
| esv3592924 | Chr.2:151812157 - 151840483 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map