Assay Details
Target Gene Details
Entrez Gene ID: | 144501 |
Gene Name: | keratin 80 |
Gene Aliases: |
KB20 |
Location: |
Chr.12:52168996-52192000 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT80 | NM_001081492.1 | NP_001074961.1 | ||
| NM_182507.2 | NP_872313.2 | |||
| XM_005268676.3 | XP_005268733.1 | |||
| AJ717743.1 | CAG30732.1 | |||
| BC065180.1 | AAH65180.1 | |||
| BU151185.1 | ||||
| BX537567.1 | CAD97784.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759899 | Chr.12:52169480 - 52391735 on Build GRCh38 | Gain |
|
| nsv832409 | Chr.12:52055382 - 52214067 on Build GRCh38 | Loss |
|
| nsv1038149 | Chr.12:52138320 - 52176448 on Build GRCh38 | Gain |
|
| esv3581594 | Chr.12:52108874 - 52176448 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map