Assay Details
Target Gene Details
Entrez Gene ID: | 79731 |
Gene Name: | asparaginyl-tRNA synthetase 2, mitochondrial (putative) |
Gene Aliases: |
DFNB94, SLM5, asnRS |
Location: |
Chr.11:78435961-78574864 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NARS2 | NM_001243251.1 | 14 | 1708 | NP_001230180.1 |
| NM_024678.5 | 14 | 2189 | NP_078954.4 | |
| XM_011545253.1 | 13 | 2155 | XP_011543555.1 | |
| XM_017018303.1 | 15 | 1773 | XP_016873792.1 | |
| XM_017018304.1 | 13 | 1674 | XP_016873793.1 | |
| AK027094.1 | BAB15655.1 | |||
| AK056980.1 | 14 | 2106 | ||
| AK225444.1 | ||||
| BC007800.2 | 14 | 1999 | AAH07800.2 | |
| BM023108.1 | 1 | 325 | ||
| DB565980.1 | 1 | 135 |
More Information
Set Membership: |
|

Genomic Map