Assay Details
Target Gene Details
Entrez Gene ID: | 85021 |
Gene Name: | RALBP1 associated Eps domain containing 1 |
Gene Aliases: |
RALBP1 |
Location: |
Chr.6:138904015-138988282 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REPS1 | NM_001128617.2 | 19 | 3152 | NP_001122089.1 |
| NM_001286611.1 | 20 | 3233 | NP_001273540.1 | |
| NM_001286612.1 | 17 | 2960 | NP_001273541.1 | |
| NM_031922.4 | 20 | 3230 | NP_114128.3 | |
| XM_005267178.4 | 19 | 3148 | XP_005267235.1 | |
| XM_017011388.1 | 19 | 2534 | XP_016866877.1 | |
| AA025663.1 | 1 | 148 | ||
| AA706721.1 | 1 | 150 | ||
| AI879474.1 | 1 | 318 | ||
| CA424106.1 | 1 | 336 |
More Information
Additional Information:
For this assay, SNP(s) [rs112317836] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map