Assay Details
Target Gene Details
Entrez Gene ID: | 5457 |
Gene Name: | POU class 4 homeobox 1 |
Gene Aliases: |
BRN3A, Oct-T1, RDC-1, brn-3A |
Location: |
Chr.13:78599092-78603560 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POU4F1 | NM_006237.3 | 2 | 3283 | NP_006228.3 |
| BM714015.1 | ||||
| L20433.1 | 2 | 3283 | AAA65605.1 | |
| X64624.1 | 1 | 3076 | CAA45907.1 |
Target Gene Details
Entrez Gene ID: | 100874222 |
Gene Name: | RNF219 antisense RNA 1 |
Gene Aliases: |
POU4F1-AS1 |
Location: |
Chr.13:78054855-78617325 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF219-AS1 | NR_047001.1 | |||
| AK095779.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv339n67 | Chr.13:78560929 - 78651758 on Build GRCh38 | Gain |
|
| nsv562410 | Chr.13:78598658 - 78603384 on Build GRCh38 | Loss |
|
| nsv826726 | Chr.13:78598212 - 78605820 on Build GRCh38 | Gain |
|
| dgv234n27 | Chr.13:78566708 - 78642283 on Build GRCh38 | Loss |
|
| nsv1097 | Chr.13:78576585 - 78609180 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map