Assay Details
Target Gene Details
Entrez Gene ID: | 2935 |
Gene Name: | G1 to S phase transition 1 |
Gene Aliases: |
551G9.2, ETF3A, GST1, eRF3a |
Location: |
Chr.16:11868128-11916662 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSPT1 | NM_001130006.1 | NP_001123478.1 | ||
| NM_001130007.1 | NP_001123479.1 | |||
| NM_002094.3 | NP_002085.2 | |||
| AK095483.1 | ||||
| AK290382.1 | ||||
| AK315267.1 | ||||
| BC009503.2 | AAH09503.2 | |||
| BT006722.1 | AAP35368.1 | |||
| X17644.1 | CAA35635.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1043909 | Chr.16:11746054 - 11913578 on Build GRCh38 | Gain |
|
| nsv571458 | Chr.16:11831151 - 11993425 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map