Assay Details
Target Gene Details
Entrez Gene ID: | 409 |
Gene Name: | arrestin beta 2 |
Gene Aliases: |
ARB2, ARR2, BARR2 |
Location: |
Chr.17:4710494-4721500 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARRB2 | NM_001257328.1 | NP_001244257.1 | ||
| NM_001257329.1 | NP_001244258.1 | |||
| NM_001257330.1 | NP_001244259.1 | |||
| NM_001257331.1 | NP_001244260.1 | |||
| NM_004313.3 | NP_004304.1 | |||
| NM_199004.1 | NP_945355.1 | |||
| NR_047516.1 | ||||
| XM_006721520.1 | XP_006721583.1 | |||
| XM_011523858.2 | XP_011522160.1 | |||
| XM_017024645.1 | XP_016880134.1 | |||
| AB209786.1 | BAD93023.1 | |||
| AF106941.1 | AAC99468.1 | |||
| AK097542.1 | BAC05094.1 | |||
| AK290951.1 | ||||
| AK297059.1 | ||||
| AK297181.1 | ||||
| BC007427.2 | AAH07427.1 | |||
| BC067368.1 | AAH67368.1 | |||
| BG723452.1 | ||||
| CR450310.1 | CAG29306.1 | |||
| CR749218.1 | CAH18075.1 | |||
| DQ664180.1 | ||||
| EU883572.1 | ||||
| Z11501.1 | CAA77577.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
| nsv833346 | Chr.17:4646256 - 4860871 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map