Assay Details
Target Gene Details
Entrez Gene ID: | 10983 |
Gene Name: | cyclin I |
Gene Aliases: |
CCNI1, CYC1, CYI |
Location: |
Chr.4:77047155-77076013 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNI | NM_006835.2 | 5 | 933 | NP_006826.1 |
| XM_011531555.1 | XP_011529857.1 | |||
| AB209766.1 | 5 | 935 | BAD93003.1 | |
| AF135162.1 | 5 | 588 | AAF43786.1 | |
| AK222931.1 | 4 | 385 | BAD96651.1 | |
| AK301111.1 | 4 | 794 | ||
| AK312632.1 | 7 | 803 | ||
| BC000420.2 | 5 | 932 | AAH00420.1 | |
| BC004975.1 | 5 | 932 | AAH04975.1 | |
| BT020019.1 | 4 | 389 | AAV38822.1 | |
| CR541783.1 | 4 | 389 | CAG46582.1 | |
| D50310.1 | 4 | 389 | BAA08849.1 |
More Information
Set Membership: |
|

Genomic Map