Assay Details
Target Gene Details
Entrez Gene ID: | 400511 |
Gene Name: | long intergenic non-protein coding RNA 1567 |
Gene Aliases: |
- |
Location: |
Chr.16:24661422-24671062 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01567 | NR_122072.1 | 3 | 2487 | |
| AK127191.1 | 3 | 2487 | BAC86880.1 |
Target Gene Details
Entrez Gene ID: | 27327 |
Gene Name: | trinucleotide repeat containing 6A |
Gene Aliases: |
CAGH26, GW1, GW182, TNRC6 |
Location: |
Chr.16:24610260-24826227 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNRC6A | XM_017023143.1 | XP_016878632.1 | ||
| XM_017023144.1 | XP_016878633.1 | |||
| XM_017023145.1 | XP_016878634.1 | |||
| XM_017023147.1 | XP_016878636.1 | |||
| XM_017023148.1 | XP_016878637.1 | |||
| XM_017023149.1 | XP_016878638.1 | |||
| XM_017023150.1 | XP_016878639.1 | |||
| XM_017023151.1 | XP_016878640.1 | |||
| XM_017023152.1 | XP_016878641.1 | |||
| U80739.1 | AAB91438.1 |
More Information
Set Membership: |
|

Genomic Map