Assay Details
Target Gene Details
Entrez Gene ID: | 9573 |
Gene Name: | growth differentiation factor 3 |
Gene Aliases: |
KFS3, MCOP7, MCOPCB6 |
Location: |
Chr.12:7689785-7695764 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GDF3 | NM_020634.1 | 2 | 536 | NP_065685.1 |
| AF110641.1 | 2 | 514 | AAM27000.1 | |
| AF263538.1 | 2 | 536 | AAF91389.1 | |
| AY358875.1 | 2 | 565 | AAQ89234.1 | |
| BC030959.1 | 2 | 548 | AAH30959.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580438 | Chr.12:7679579 - 7712391 on Build GRCh38 | Gain |
|
| esv2422393 | Chr.12:7682970 - 7834994 on Build GRCh38 | Duplication |
|
| nsv557274 | Chr.12:7673709 - 7712380 on Build GRCh38 | Gain |
|
| dgv6e196 | Chr.12:7670828 - 7920436 on Build GRCh38 | Duplication |
|
| nsv557273 | Chr.12:7621432 - 7731755 on Build GRCh38 | Loss |
|
| dgv5e196 | Chr.12:7621924 - 7954221 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map