Assay Details
Target Gene Details
Entrez Gene ID: | 9014 |
Gene Name: | TATA-box binding protein associated factor, RNA polymerase I subunit B |
Gene Aliases: |
MGC:9349, RAF1B, RAFI63, SL1, TAFI63 |
Location: |
Chr.2:9843442-9934416 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAF1B | NM_001318976.1 | 7 | 686 | NP_001305905.1 |
| NM_001318977.1 | 6 | 556 | NP_001305906.1 | |
| NM_005680.2 | 6 | 556 | NP_005671.2 | |
| XM_011510410.2 | 6 | 497 | XP_011508712.1 | |
| XM_011510411.1 | 7 | 605 | XP_011508713.1 | |
| AK026521.1 | 6 | 473 | ||
| AK295402.1 | 7 | 605 | ||
| AK308070.1 | 6 | 505 | ||
| AK309751.1 | 6 | 539 | ||
| AK315098.1 | 6 | 488 | ||
| BC018137.2 | 6 | 555 | AAH18137.1 | |
| L39061.1 | 6 | 558 | AAA62863.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003026 | Chr.2:9714810 - 9872387 on Build GRCh38 | Gain |
|
| nsv580960 | Chr.2:9853322 - 9926565 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115645168] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map