Assay Details
Target Gene Details
Entrez Gene ID: | 1448 |
Gene Name: | casein kappa |
Gene Aliases: |
CNS10, CSN10, CSNK, KCA |
Location: |
Chr.4:70238370-70251432 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSN3 | NM_005212.2 | NP_005203.2 | ||
| XM_011531620.2 | XP_011529922.1 | |||
| XM_017007761.1 | XP_016863250.1 | |||
| BC010935.1 | AAH10935.1 | |||
| BG191674.1 | ||||
| M73628.1 | AAA59456.1 | |||
| X66417.1 | CAA47048.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv998828 | Chr.4:70198151 - 70302146 on Build GRCh38 | Gain |
|
| nsv594646 | Chr.4:70240300 - 70271806 on Build GRCh38 | Gain |
|
| nsv1012495 | Chr.4:69979953 - 70947088 on Build GRCh38 | Gain |
|
| nsv829963 | Chr.4:70137544 - 70289392 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map