Assay Details
Target Gene Details
Entrez Gene ID: | 917 |
Gene Name: | CD3g molecule |
Gene Aliases: |
CD3-GAMMA, IMD17, T3G |
Location: |
Chr.11:118344307-118353782 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD3G | NM_000073.2 | 3 | 307 | NP_000064.1 |
| XM_005271724.3 | 3 | 344 | XP_005271781.1 | |
| XM_006718941.2 | 3 | 295 | XP_006719004.1 | |
| AK223314.1 | 3 | 260 | BAD97034.1 | |
| AK299997.1 | 2 | 999 | ||
| AK310578.1 | 4 | 435 | ||
| AK310622.1 | 4 | 465 | ||
| AK313966.1 | 3 | 295 | ||
| AK315895.1 | 3 | 251 | ||
| BC113830.1 | 3 | 253 | AAI13831.1 | |
| X04145.1 | 3 | 264 | CAA27764.1 | |
| X60492.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv820176 | Chr.11:118347470 - 118349870 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map