Assay Details
Target Gene Details
Entrez Gene ID: | 347733 |
Gene Name: | tubulin beta 2B class IIb |
Gene Aliases: |
PMGYSA, bA506K6.1 |
Location: |
Chr.6:3224261-3227734 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBB2B | NM_178012.4 | 4 | 1641 | NP_821080.1 |
| AF035316.1 | 1 | 1143 | AAB88188.1 | |
| AK289433.1 | 4 | 1562 | ||
| AK297568.1 | 3 | 1287 | ||
| AL050056.1 | 1 | 1090 | CAB43252.1 | |
| BC001352.2 | 4 | 1641 | AAH01352.1 | |
| BC063610.1 | 4 | 1544 | AAH63610.1 | |
| BX419454.2 | 1 | 240 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv965676 | Chr.6:3224204 - 3226178 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map