Assay Details
Target Gene Details
Entrez Gene ID: | 55081 |
Gene Name: | intraflagellar transport 57 |
Gene Aliases: |
ESRRBL1, HIPPI, MHS4R2 |
Location: |
Chr.3:108160812-108222570 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFT57 | NM_018010.3 | 11 | 3074 | NP_060480.1 |
| AF139576.1 | 11 | 2904 | AAK13588.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011158 | Chr.3:107746970 - 108629120 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map