Assay Details
Target Gene Details
Entrez Gene ID: | 4739 |
Gene Name: | neural precursor cell expressed, developmentally down-regulated 9 |
Gene Aliases: |
CAS-L, CAS2, CASL, CASS2, HEF1 |
Location: |
Chr.6:11183298-11382348 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEDD9 | NM_001142393.1 | NP_001135865.1 | ||
| NM_001271033.1 | NP_001257962.1 | |||
| NM_006403.3 | NP_006394.1 | |||
| NR_073131.1 | ||||
| AK292682.1 | ||||
| BC040207.1 | AAH40207.1 | |||
| BX648041.1 | ||||
| L43821.1 | AAA98770.1 | |||
| U64317.1 | AAB53696.1 |
More Information
Set Membership: |
|

Genomic Map