Assay Details
Target Gene Details
Entrez Gene ID: | 56922 |
Gene Name: | methylcrotonoyl-CoA carboxylase 1 |
Gene Aliases: |
MCC-B, MCCA |
Location: |
Chr.3:183015218-183099587 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCCC1 | NM_001293273.1 | 14 | 1698 | NP_001280202.1 |
| NM_020166.4 | 16 | 1957 | NP_064551.3 | |
| NR_120639.1 | 13 | 1674 | ||
| NR_120640.1 | 16 | 2357 | ||
| XM_006713702.1 | 15 | 1763 | XP_006713765.1 | |
| XM_011512992.2 | 16 | 2555 | XP_011511294.1 | |
| XM_017006874.1 | 15 | 2322 | XP_016862363.1 | |
| XM_017006875.1 | 14 | 1559 | XP_016862364.1 | |
| AB029826.1 | 16 | 1942 | BAA99407.1 | |
| AB209737.1 | 16 | 1858 | BAD92974.1 | |
| AF297332.1 | 16 | 1976 | AAK67986.1 | |
| AF310339.1 | 16 | 1942 | AAG50245.1 | |
| AF310972.1 | 16 | 1943 | AAG53095.1 | |
| AK023051.1 | 16 | 1933 | BAB14377.1 | |
| AK098411.1 | 6 | 643 | ||
| AK294882.1 | 13 | 1605 | ||
| AK295293.1 | 15 | 1778 | ||
| AL442091.1 | 16 | 1876 | ||
| BC004187.2 | 16 | 1942 | AAH04187.1 | |
| BC004214.2 | 16 | 1942 | AAH04214.1 | |
| CR749608.1 | 14 | 1570 | CAH18403.1 | |
| KU178727.1 | 16 | 1810 | ||
| KU178728.1 | 5 | 381 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv592654 | Chr.3:182938822 - 183025479 on Build GRCh38 | Loss |
|
| nsv482968 | Chr.3:182869307 - 183035390 on Build GRCh38 | Gain |
|
| nsv508968 | Chr.3:182974407 - 183063558 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map