Assay Details
Target Gene Details
Entrez Gene ID: | 4077 |
Gene Name: | NBR1, autophagy cargo receptor |
Gene Aliases: |
1A1-3B, IAI3B, M17S2, MIG19 |
Location: |
Chr.17:43170471-43211689 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NBR1 | NM_031862.3 | NP_114068.1 | ||
| XM_011524813.1 | 1 | 321 | XP_011523115.1 | |
| XM_017024642.1 | 1 | 294 | XP_016880131.1 | |
| XM_017024643.1 | 1 | 321 | XP_016880132.1 | |
| XM_017024644.1 | XP_016880133.1 | |||
| AF227189.1 | 1 | 329 | AAF74119.1 | |
| D30756.1 | ||||
| DB081342.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2672913 | Chr.17:42964983 - 43322347 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map